Learn More
Dichlorobis(tri-o-tolylphosphine)palladium(II), 95%
CAS: 40691-33-6 | C42H42Cl2P2Pd | 786.06 g/mol
$113.70 - $113.70
Chemical Identifiers
| CAS | 40691-33-6 |
|---|---|
| Molecular Formula | C42H42Cl2P2Pd |
| Molecular Weight (g/mol) | 786.06 |
| MDL Number | MFCD00274659 |
| InChI Key | GMLMZLGOMMIIMA-UHFFFAOYSA-L |
| Synonym | palladium,dichlorobis tris 2-methylphenyl phosphine |
| PubChem CID | 90656943 |
| IUPAC Name | palladium;tris(2-methylphenyl)phosphane;dichloride |
| SMILES | Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1 |
Description
Dichlorobis(tri-o-tolylphosphine)palladium(II) is used in reaction of tributyltin enolates, prepared in situ from tributyltin methoxide and enol acetates, with aryl bromides, coupling reaction of aryl bromides with vinylic acetates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 40691-33-6 | |
| 786.06 | |
| GMLMZLGOMMIIMA-UHFFFAOYSA-L | |
| 90656943 | |
| Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1 |
| C42H42Cl2P2Pd | |
| MFCD00274659 | |
| palladium,dichlorobis tris 2-methylphenyl phosphine | |
| palladium;tris(2-methylphenyl)phosphane;dichloride |
Specifications
| 40691-33-6 | |
| C42H42Cl2P2Pd | |
| 1 g | |
| palladium,dichlorobis tris 2-methylphenyl phosphine | |
| GMLMZLGOMMIIMA-UHFFFAOYSA-L | |
| palladium;tris(2-methylphenyl)phosphane;dichloride | |
| 90656943 | |
| 98% | |
| Dichlorobis (tri-o-tolylphosphine)palladium (II) |
| Yellow | |
| MFCD00274659 | |
| Air and moisture sensitive | |
| Soluble in ethanol,benzene,and ethereal solvents. | |
| Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1 | |
| 786.06 | |
| 786.08 | |
| Powder |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only