accessibility menu, dialog, popup

UserName

CAS RN 40691-33-6

IUPAC Name: dichloropalladiumbis(ylium); bis(tris(2-methylphenyl)phosphane)
Synonyms: dichloropalladiumbis(ylium); bis(tris(o-tolyl)phosphine)
Molecular Weight (g/mol): 786.06
Molecular Formula: C42H42Cl2P2Pd
InChi Key: GMLMZLGOMMIIMA-UHFFFAOYSA-L
SMILES: Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (4)
  • (1)
Quantity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)