missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II), Pd 13% min

Catalog No. p-7059603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4497703 1 g
AA4497706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4497703 Supplier Thermo Scientific Chemicals Supplier No. 04497703
Only null left

CAS: 205319-06-8 | C36H28Cl2OP2Pd | 715.89 g/mol

Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II) is used as a catalyst for cross-coupling reaction of 2-brom-1,3-dienes to prepare high stereoselectivity of conjugated Z,E dienes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 205319-06-8
Molecular Formula C36H28Cl2OP2Pd
Molecular Weight (g/mol) 715.89
MDL Number MFCD09953446
InChI Key QZIOEBWDZNUTLZ-UHFFFAOYSA-L
Synonym dichloro bis diphenylphosphinophenyl ether palladium ii, dichloro bis 2-diphenylphosphino phenyl ether palladium ii, bis diphenylphosphinophenyl ether palladium ii dichloride, dichloro bis diphenylphosphinophenyl ether palladium ii , pd, oxydi 2,1-phenylene bis diphenylphosphane-dichloropalladium 1/1, palladium chloride; 2-2-diphenylphosphanyl phenoxy phenyl diphenylphosphane
PubChem CID 11377567
IUPAC Name dichloropalladium;[2-(2-diphenylphosphanylphenoxy)phenyl]-diphenylphosphane
SMILES Cl[Pd++]Cl.O(C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 715.88
Concentration or Composition (by Analyte or Components) Pd 14.8%
Chemical Name or Material Dichloro[bis (diphenylphosphinophenyl)ether]palladium (II)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only