missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloramine T 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 79°C |
| Color | Yellow |
| Quantity | 500 g |
| UN Number | 1479 |
| Formula Weight | 240.10 |
| Percent Purity | ≥96.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dichloramine T |
Safety and Handling
| TSCA | Yes |