accessibility menu, dialog, popup

UserName

CH3SOONClCl

CAS RN 473-34-7

IUPAC Name: N,N-dichloro-4-methylbenzene-1-sulfonamide
Synonyms: N,N-dichloro-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 240.1
Molecular Formula: C7H7Cl2NO2S
InChi Key: ARGDYOIRHYLIMT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N(Cl)Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results