missing translation for 'onlineSavingsMsg'
Learn More

Dichloramine T 96.0+%, TCI America™

Catalog No. D031825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D031825G 25 g
D0318500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D031825G Supplier TCI America Supplier No. D031825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 473-34-7
Molecular Formula C7H7Cl2NO2S
Molecular Weight (g/mol) 240.098
MDL Number MFCD00058929
InChI Key ARGDYOIRHYLIMT-UHFFFAOYSA-N
Synonym N,N-Dichloro-p-toluenesulfonamide, p-Toluenesulfondichloramide
PubChem CID 68056
IUPAC Name N,N-dichloro-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N(Cl)Cl

Specifications

Melting Point 79°C
Color Yellow
Quantity 25 g
UN Number 1479
Formula Weight 240.10
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dichloramine T