missing translation for 'onlineSavingsMsg'
Learn More

Dicamba 98.0+%, TCI America™

Supplier:  TCI America D4800200MG

Catalog No. D4800200MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dicamba
115°C
C8H6Cl2O3
200 mg
IWEDIXLBFLAXBO-UHFFFAOYSA-N
3,6-dichloro-2-methoxybenzoic acid
3030
221.03
Crystalline Powder
1918-00-9
White
MFCD00055283
dicamba, mdba, mediben, banvel, dianat, 3,6-dichloro-o-anisic acid, banlen, brush buster, banvel herbicide, banvel cst
COC1=C(C=CC(=C1C(=O)O)Cl)Cl
221.033
CHEBI:81856
≥98.0% (GC,T)

Chemical Identifiers

1918-00-9
221.033
IWEDIXLBFLAXBO-UHFFFAOYSA-N
3030
3,6-dichloro-2-methoxybenzoic acid
C8H6Cl2O3
MFCD00055283
dicamba, mdba, mediben, banvel, dianat, 3,6-dichloro-o-anisic acid, banlen, brush buster, banvel herbicide, banvel cst
CHEBI:81856
COC1=C(C=CC(=C1C(=O)O)Cl)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3053

RTECSNumber : DG7525000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions