missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Succinate 99.0+%, TCI America™

Supplier:  TCI America S010225ML

 View more versions of this product

Catalog No. S010225ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dibutyl Succinate
-29°C
274°C
MFCD00040495
dibutyl succinate, tabutrex, succinic acid di-n-butyl ester, di-n-butyl succinate, tabatrex, dibutylsuccinate, butyl butanedioate, butanedioic acid, dibutyl ester, succinic acid dibutyl ester, dnbs
CCCCOC(=O)CCC(=O)OCCCC
230.304
CHEBI:82263
≥99.0% (GC)
141-03-7
Colorless
C12H22O4
25 mL
YUXIBTJKHLUKBD-UHFFFAOYSA-N
dibutyl butanedioate
8830
230.30
Liquid

Chemical Identifiers

141-03-7
230.304
YUXIBTJKHLUKBD-UHFFFAOYSA-N
8830
dibutyl butanedioate
C12H22O4
MFCD00040495
dibutyl succinate, tabutrex, succinic acid di-n-butyl ester, di-n-butyl succinate, tabatrex, dibutylsuccinate, butyl butanedioate, butanedioic acid, dibutyl ester, succinic acid dibutyl ester, dnbs
CHEBI:82263
CCCCOC(=O)CCC(=O)OCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0848

RTECSNumber : WM6250000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions