missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibutyl Succinate 99.0+%, TCI America™
$78.93 - $78.93
Chemical Identifiers
| CAS | 141-03-7 |
|---|---|
| Molecular Formula | C12H22O4 |
| Molecular Weight (g/mol) | 230.304 |
| MDL Number | MFCD00040495 |
| InChI Key | YUXIBTJKHLUKBD-UHFFFAOYSA-N |
| Synonym | dibutyl succinate, tabutrex, succinic acid di-n-butyl ester, di-n-butyl succinate, tabatrex, dibutylsuccinate, butyl butanedioate, butanedioic acid, dibutyl ester, succinic acid dibutyl ester, dnbs |
| PubChem CID | 8830 |
| ChEBI | CHEBI:82263 |
| IUPAC Name | dibutyl butanedioate |
| SMILES | CCCCOC(=O)CCC(=O)OCCCC |
Chemical Identifiers
| 141-03-7 | |
| 230.304 | |
| YUXIBTJKHLUKBD-UHFFFAOYSA-N | |
| 8830 | |
| dibutyl butanedioate |
| C12H22O4 | |
| MFCD00040495 | |
| dibutyl succinate, tabutrex, succinic acid di-n-butyl ester, di-n-butyl succinate, tabatrex, dibutylsuccinate, butyl butanedioate, butanedioic acid, dibutyl ester, succinic acid dibutyl ester, dnbs | |
| CHEBI:82263 | |
| CCCCOC(=O)CCC(=O)OCCCC |
Specifications
| 141-03-7 | |
| Colorless | |
| C12H22O4 | |
| 25 mL | |
| YUXIBTJKHLUKBD-UHFFFAOYSA-N | |
| dibutyl butanedioate | |
| 8830 | |
| 230.30 | |
| Liquid |
| -29°C | |
| 274°C | |
| MFCD00040495 | |
| dibutyl succinate, tabutrex, succinic acid di-n-butyl ester, di-n-butyl succinate, tabatrex, dibutylsuccinate, butyl butanedioate, butanedioic acid, dibutyl ester, succinic acid dibutyl ester, dnbs | |
| CCCCOC(=O)CCC(=O)OCCCC | |
| 230.304 | |
| CHEBI:82263 | |
| ≥99.0% (GC) | |
| Dibutyl Succinate |
Safety and Handling
EINECSNumber : (2)-0848
RTECSNumber : WM6250000
TSCA : Yes