accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 141-03-7

IUPAC Name: 1,4-dibutyl butanedioate
Synonyms: butanedioic acid, dibutyl ester
Molecular Weight (g/mol): 230.3
Molecular Formula: C12H22O4
InChi Key: YUXIBTJKHLUKBD-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CCC(=O)OCCCC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)