missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Phthalate, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor DI1154LTGL

 View more versions of this product

Catalog No. 18610171


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

84-74-2
340°C
4 L
Reagent
1
Steel Can
0.98
Liquid

Chemical Identifiers

84-74-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Not for pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.