missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Phthalate, 98%, Spectrum™ Chemical

Catalog No. p-7165499
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610172 500 mL Steel Can
18610171 4 L Steel Can
18610169 20 L Steel Can
18610170 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610172 Supplier Spectrum Chemical Mfg Cor Supplier No. DI115500MLGL
Only null left

Chemical Identifiers

CAS 84-74-2

Specifications

CAS Max % 1
Boiling Point 340°C
Packaging Steel Can
Quantity 500 mL
Percent Purity 0.98
Grade Reagent
Physical Form Liquid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Not for pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.