missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Methylenebis(thioglycolate) 95.0+%, TCI America™

Catalog No. M113725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M113725G 25 g
M1137500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M113725G Supplier TCI America Supplier No. M113725G
Only null left

Chemical Identifiers

CAS 14338-82-0
Molecular Formula C13H24O4S2
Molecular Weight (g/mol) 308.451
MDL Number MFCD00027216
InChI Key QCWRDNFBMVSDEY-UHFFFAOYSA-N
Synonym Methylenebis(thioglycolic Acid Dibutyl Ester)
PubChem CID 84361
IUPAC Name butyl 2-[(2-butoxy-2-oxoethyl)sulfanylmethylsulfanyl]acetate
SMILES CCCCOC(=O)CSCSCC(=O)OCCCC

Specifications

Color Colorless
Boiling Point 178°C
Quantity 25 g
Formula Weight 308.45
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyl Methylenebis(thioglycolate)
EINECSNumber (2)-1401
TSCA Yes