missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibutyl Disulfide 95.0+%, TCI America™
Supplier: TCI America D0224500ML
Specifications
| Dibutyl Disulfide | |
| Yellow | |
| C8H18S2 | |
| 500 mL | |
| CUDSBWGCGSUXDB-UHFFFAOYSA-N | |
| 1-(butyldisulfanyl)butane | |
| 12386 | |
| ≥95.0% (GC) |
| 629-45-8 | |
| 88°C | |
| MFCD00009467 | |
| dibutyl disulfide, butyl disulfide, disulfide, dibutyl, n-butyl disulfide, di-n-butyl disulfide, dibutyl disulphide, 5,6-dithiadecane, 1-butyldisulfanyl butane, unii-z77ef35c0q, 1-butyldisulfanyl-butane | |
| CCCCSSCCCC | |
| 178.35 | |
| 178.35 | |
| Liquid |
Chemical Identifiers
| 629-45-8 | |
| 178.35 | |
| CUDSBWGCGSUXDB-UHFFFAOYSA-N | |
| 12386 | |
| CCCCSSCCCC |
| C8H18S2 | |
| MFCD00009467 | |
| dibutyl disulfide, butyl disulfide, disulfide, dibutyl, n-butyl disulfide, di-n-butyl disulfide, dibutyl disulphide, 5,6-dithiadecane, 1-butyldisulfanyl butane, unii-z77ef35c0q, 1-butyldisulfanyl-butane | |
| 1-(butyldisulfanyl)butane |
Safety and Handling
EINECSNumber : (2)-0477
TSCA : Yes