Learn More
Dibromomethane, 99%
CAS: 74-95-3 | CH2Br2 | 173.835 g/mol
Supplier: Thermo Scientific Chemicals A104560E
Description
Dibromomethane is used as solvent in organic synthesis. It acts as an intermediate in the manufacture of specialty chemicals, agrochemicals and pharmaceuticals. It is useful as extractant and utilized for the determination of 5-nitroimidazoles (5-NDZ) in environmental waters. It is involved in the convertion of catechols to their methylenedioxy derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDibromomethane is used as solvent in organic synthesis. It acts as an intermediate in the manufacture of specialty chemicals, agrochemicals and pharmaceuticals. It is useful as extractant and utilized for the determination of 5-nitroimidazoles (5-NDZ) in environmental waters. It is involved in the convertion of catechols to their methylenedioxy derivatives.
Solubility
Miscible with water, chloroform, acetone, ether and alcohol.
Notes
Store in cool place. Incompatible with strong oxidizing agents, aluminum and magnesium.
Specifications
| Dibromomethane | |
| 74-95-3 | |
| 2.495 | |
| CH2Br2 | |
| MFCD00000168 | |
| 2664 | |
| 14,6061 | |
| Miscible with water,chloroform,acetone,ether and alcohol. | |
| C(Br)Br | |
| 173.835 | |
| CHEBI:47077 | |
| 99% |
| Stabilized with 50ppm BHT | |
| -53°C | |
| 96°C to 98°C | |
| 1.541 | |
| 2500 g | |
| 969143 | |
| methylene bromide, methane, dibromo, methylene dibromide, dibromomethylene, rcra waste number u068, dibrommethan, methylenbromid, dibromo-methane, ccris 939, rcra waste no. u068 | |
| FJBFPHVGVWTDIP-UHFFFAOYSA-N | |
| dibromomethane | |
| 3024 | |
| 173.85 |
Chemical Identifiers
| 74-95-3 | |
| 173.835 | |
| FJBFPHVGVWTDIP-UHFFFAOYSA-N | |
| 3024 | |
| dibromomethane |
| CH2Br2 | |
| MFCD00000168 | |
| methylene bromide, methane, dibromo, methylene dibromide, dibromomethylene, rcra waste number u068, dibrommethan, methylenbromid, dibromo-methane, ccris 939, rcra waste no. u068 | |
| CHEBI:47077 | |
| C(Br)Br |
Safety and Handling
GHS H Statement
H332
Harmful if inhaled.
P261-P271-P304+P340-P312
H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DIBROMOMETHANE
EINECSNumber : 200-824-2
RTECSNumber : PA7350000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only