missing translation for 'onlineSavingsMsg'
Learn More

Dibromomethane, 99%

Catalog No. p-7080719
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1045622 100 g
AAA1045636 500 g
AAA104560E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1045622 Supplier Thermo Scientific Chemicals Supplier No. A1045622
Only null left

CAS: 74-95-3 | CH2Br2 | 173.835 g/mol

Dibromomethane is used as solvent in organic synthesis. It acts as an intermediate in the manufacture of specialty chemicals, agrochemicals and pharmaceuticals. It is useful as extractant and utilized for the determination of 5-nitroimidazoles (5-NDZ) in environmental waters. It is involved in the convertion of catechols to their methylenedioxy derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibromomethane is used as solvent in organic synthesis. It acts as an intermediate in the manufacture of specialty chemicals, agrochemicals and pharmaceuticals. It is useful as extractant and utilized for the determination of 5-nitroimidazoles (5-NDZ) in environmental waters. It is involved in the convertion of catechols to their methylenedioxy derivatives.

Solubility
Miscible with water, chloroform, acetone, ether and alcohol.

Notes
Store in cool place. Incompatible with strong oxidizing agents, aluminum and magnesium.

Chemical Identifiers

CAS 74-95-3
Molecular Formula CH2Br2
Molecular Weight (g/mol) 173.835
MDL Number MFCD00000168
InChI Key FJBFPHVGVWTDIP-UHFFFAOYSA-N
Synonym methylene bromide, methane, dibromo, methylene dibromide, dibromomethylene, rcra waste number u068, dibrommethan, methylenbromid, dibromo-methane, ccris 939, rcra waste no. u068
PubChem CID 3024
ChEBI CHEBI:47077
IUPAC Name dibromomethane
SMILES C(Br)Br

Specifications

Melting Point -53°C
Density 2.495
Boiling Point 96°C to 98°C
Refractive Index 1.541
Quantity 100 g
UN Number UN2664
Beilstein 969143
Merck Index 14,6061
Solubility Information Miscible with water,chloroform,acetone,ether and alcohol.
Formula Weight 173.85
Percent Purity 99%
Chemical Name or Material Dibromomethane, Stabilized with 50ppm BHT
Show More Show Less
Hazard Category H332
Hazard Statement GHS H Statement
H332
Harmful if inhaled.
Precautionary Statement P261-P271-P304+P340-P312
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DIBROMOMETHANE
EINECSNumber 200-824-2
RTECSNumber PA7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only