accessibility menu, dialog, popup

UserName

BrBr

CAS RN 74-95-3

IUPAC Name: dibromomethane
Synonyms: dibromomethane methylene bromide ion (1+) methylenbromid ccris 939 dibromomethane (substance) rcra waste number U068 methylene bromide methylene dibromide 82br-labeled methylene bromide 80br-labeled methylene bromide
Molecular Weight (g/mol): 173.84
Molecular Formula: CH2Br2
InChi Key: FJBFPHVGVWTDIP-UHFFFAOYSA-N
SMILES: BrCBr
Boiling Point 
  • (3)
  • (4)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (1)
  • (1)
  • (2)
  • (7)
  • (3)
  • (4)
  • (1)
  • (2)
  • (4)
  • (5)
  • (1)
  • (2)
  • (1)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (7)
  • (1)