missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibromomalonamide 98.0+%, TCI America™
$59.65 - $193.86
Chemical Identifiers
| CAS | 73003-80-2 |
|---|---|
| Molecular Formula | C3H4Br2N2O2 |
| Molecular Weight (g/mol) | 259.89 |
| MDL Number | MFCD00014803 |
| InChI Key | SWHQVMGRXIYDSF-UHFFFAOYSA-N |
| Synonym | dibromomalonamide, 2,2-dibromomalonamide, dibromopropanediamide, propanediamide, 2,2-dibromo, dibromo malonamide-13c3, dibromo malonamide, dibromomalonic diamide, dibromomalonic acid diamide, propanediamide,2,2-dibromo, 2,2-bis bromanyl propanediamide |
| PubChem CID | 175375 |
| IUPAC Name | dibromopropanediamide |
| SMILES | NC(=O)C(Br)(Br)C(N)=O |
Chemical Identifiers
| 73003-80-2 | |
| 259.89 | |
| SWHQVMGRXIYDSF-UHFFFAOYSA-N | |
| 175375 | |
| NC(=O)C(Br)(Br)C(N)=O |
| C3H4Br2N2O2 | |
| MFCD00014803 | |
| dibromomalonamide, 2,2-dibromomalonamide, dibromopropanediamide, propanediamide, 2,2-dibromo, dibromo malonamide-13c3, dibromo malonamide, dibromomalonic diamide, dibromomalonic acid diamide, propanediamide,2,2-dibromo, 2,2-bis bromanyl propanediamide | |
| dibromopropanediamide |
Specifications
| 73003-80-2 | |
| White | |
| MFCD00014803 | |
| dibromomalonamide, 2,2-dibromomalonamide, dibromopropanediamide, propanediamide, 2,2-dibromo, dibromo malonamide-13c3, dibromo malonamide, dibromomalonic diamide, dibromomalonic acid diamide, propanediamide,2,2-dibromo, 2,2-bis bromanyl propanediamide | |
| NC(=O)C(Br)(Br)C(N)=O | |
| 259.89 | |
| 259.89 | |
| Crystalline Powder |
| 204°C | |
| C3H4Br2N2O2 | |
| 5 g | |
| SWHQVMGRXIYDSF-UHFFFAOYSA-N | |
| dibromopropanediamide | |
| 175375 | |
| ≥98.0% (N) | |
| Dibromomalonamide |
Safety and Handling
TSCA : Yes