missing translation for 'onlineSavingsMsg'
Learn More

Dibromomalonamide 98.0+%, TCI America™

Supplier:  TCI America D427225G

 View more versions of this product

Catalog No. D427225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dibromomalonamide
204°C
C3H4Br2N2O2
25 g
SWHQVMGRXIYDSF-UHFFFAOYSA-N
dibromopropanediamide
175375
≥98.0% (N)
73003-80-2
White
MFCD00014803
dibromomalonamide, 2,2-dibromomalonamide, dibromopropanediamide, propanediamide, 2,2-dibromo, dibromo malonamide-13c3, dibromo malonamide, dibromomalonic diamide, dibromomalonic acid diamide, propanediamide,2,2-dibromo, 2,2-bis bromanyl propanediamide
NC(=O)C(Br)(Br)C(N)=O
259.89
259.89
Crystalline Powder

Chemical Identifiers

73003-80-2
259.89
SWHQVMGRXIYDSF-UHFFFAOYSA-N
175375
NC(=O)C(Br)(Br)C(N)=O
C3H4Br2N2O2
MFCD00014803
dibromomalonamide, 2,2-dibromomalonamide, dibromopropanediamide, propanediamide, 2,2-dibromo, dibromo malonamide-13c3, dibromo malonamide, dibromomalonic diamide, dibromomalonic acid diamide, propanediamide,2,2-dibromo, 2,2-bis bromanyl propanediamide
dibromopropanediamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions