accessibility menu, dialog, popup

UserName

H2NOBrBrNH2O

CAS RN 73003-80-2

IUPAC Name: dibromopropanediamide
Synonyms: dibromopropanediamide
Molecular Weight (g/mol): 259.89
Molecular Formula: C3H4Br2N2O2
InChi Key: SWHQVMGRXIYDSF-UHFFFAOYSA-N
SMILES: NC(=O)C(Br)(Br)C(N)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)