missing translation for 'onlineSavingsMsg'
Learn More

Dibenzylamine 97.0+%, TCI America™

Catalog No. p-7126060
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D015225G 25 g
D0152500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D015225G Supplier TCI America Supplier No. D015225G
Only null left

Chemical Identifiers

CAS 103-49-1
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.28
MDL Number MFCD00004770
InChI Key BWLUMTFWVZZZND-UHFFFAOYSA-N
Synonym dibenzylamine, benzenemethanamine, n-phenylmethyl, n-benzylbenzylamine, bibenzylamine, dibenzyl-amine, dibenzyl amine, bisbenzylamine, n,n-dibenzylamine, n-benzylaminomethyl benzene, unii-3g0yfx01c6
PubChem CID 7656
IUPAC Name dibenzylamine
SMILES C(NCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -26°C
Color Yellow
Boiling Point 270°C
Quantity 25 g
Formula Weight 197.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibenzylamine
EINECSNumber (3)-0315&(9)-0738
TSCA Yes