accessibility menu, dialog, popup

UserName

CAS RN 103-49-1

IUPAC Name: dibenzylamine
Synonyms: dibenzyl amine
Molecular Weight (g/mol): 197.28
Molecular Formula: C14H15N
InChi Key: BWLUMTFWVZZZND-UHFFFAOYSA-N
SMILES: C(NCC1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)