missing translation for 'onlineSavingsMsg'
Learn More

Dibenzylamine, 98%

Catalog No. AAA1155430
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1155430 250 g
AAA115540B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1155430 Supplier Thermo Scientific Chemicals Supplier No. A1155430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 103-49-1 | C14H15N | 197.28 g/mol

Dibenzylamine is used to prepare rubber accelerator for the vulcanization process and reaction-stoppers. Dibenzylamine type accelerators reduce nitrosamine production in the compounding process. It is also used to produce dibenzyl-nitroso-amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzylamine is used to prepare rubber accelerator for the vulcanization process and reaction-stoppers. Dibenzylamine type accelerators reduce nitrosamine production in the compounding process. It is also used to produce dibenzyl-nitroso-amine.

Solubility
Soluble in water(0.4g/L).

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 103-49-1
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.28
MDL Number MFCD00004770
InChI Key BWLUMTFWVZZZND-UHFFFAOYSA-N
Synonym dibenzylamine, benzenemethanamine, n-phenylmethyl, n-benzylbenzylamine, bibenzylamine, dibenzyl-amine, dibenzyl amine, bisbenzylamine, n,n-dibenzylamine, n-benzylaminomethyl benzene, unii-3g0yfx01c6
PubChem CID 7656
SMILES C(NCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -26°C
Density 1.026
pH 8.9
Boiling Point 300°C
Flash Point 143°C (289°F)
Odor Weak
Refractive Index 1.5745
Quantity 250 g
UN Number UN2735
Beilstein 909664
Merck Index 14,3011
Solubility Information Soluble in water(0.4g/L).
IUPAC Name N-benzyl-1-phenylmethanamine
Formula Weight 197.28
Percent Purity 98%
Chemical Name or Material Dibenzylamine
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 203-117-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only