missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Ether 95.0+%, TCI America™

Catalog No. p-7118434
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
B041825ML 25 mL
B0418500ML 500 mL
2 options

Catalog No. B041825ML

Supplier: TCI America B041825ML

Only null left

Chemical Identifiers

CAS 103-50-4
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.265
MDL Number MFCD00004780
InChI Key MHDVGSVTJDSBDK-UHFFFAOYSA-N
Synonym dibenzyl ether, benzyl ether, benzyl oxide, dibenzylether, oxybis methylene dibenzene, plastikator ba, ba plasticizer, benzylether, ether, dibenzyl, plasticator ba
PubChem CID 7657
ChEBI CHEBI:87411
IUPAC Name phenylmethoxymethylbenzene
SMILES C1=CC=C(C=C1)COCC2=CC=CC=C2

Specifications

Melting Point 4°C
Color Yellow
Boiling Point 298°C
Quantity 25 mL
UN Number 3082
Formula Weight 198.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibenzyl Ether
EINECSNumber (3)-1082
RTECSNumber DQ6125000
TSCA Yes