accessibility menu, dialog, popup

UserName

CAS RN 103-50-4

IUPAC Name: [(benzyloxy)methyl]benzene
Synonyms: dibenzyl ether
Molecular Weight (g/mol): 198.27
Molecular Formula: C14H14O
InChi Key: MHDVGSVTJDSBDK-UHFFFAOYSA-N
SMILES: C(OCC1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (5)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)