missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Ether 95.0+%, TCI America™

Catalog No. B0418500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0418500ML 500 mL
B041825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0418500ML Supplier TCI America Supplier No. B0418500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103-50-4
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.265
MDL Number MFCD00004780
InChI Key MHDVGSVTJDSBDK-UHFFFAOYSA-N
Synonym dibenzyl ether, benzyl ether, benzyl oxide, dibenzylether, oxybis methylene dibenzene, plastikator ba, ba plasticizer, benzylether, ether, dibenzyl, plasticator ba
PubChem CID 7657
ChEBI CHEBI:87411
IUPAC Name phenylmethoxymethylbenzene
SMILES C1=CC=C(C=C1)COCC2=CC=CC=C2

Specifications

Melting Point 4°C
Color Yellow
Boiling Point 298°C
Quantity 500 mL
UN Number 3082
Formula Weight 198.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibenzyl Ether
EINECSNumber (3)-1082
RTECSNumber DQ6125000
TSCA Yes