missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl ether, 98+%

Catalog No. AAA1844730
Change view
Click to view available options
Quantity:
250 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1844730 250 g
AAA1844736 500 g
AAA184470E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1844730 Supplier Thermo Scientific Chemicals Supplier No. A1844730
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 103-50-4 | C14H14O | 198.265 g/mol

Benzyl ether is used as an effective GFP friendly tissue clearing medium for mouse brains. It is also used in a study to develop a detailed protocol for performing 3D imaging of solvent-cleared organs and its application to various microscopy techniques. It is also employed as a plasticizer for nitrocellulose and synthetic rubber, a solvent in perfumery, and a flavoring agent in chewing gums and baked goods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl ether is used as an effective GFP friendly tissue clearing medium for mouse brains. It is also used in a study to develop a detailed protocol for performing 3D imaging of solvent-cleared organs and its application to various microscopy techniques. It is also employed as a plasticizer for nitrocellulose and synthetic rubber, a solvent in perfumery, and a flavoring agent in chewing gums and baked goods.

Solubility
Not miscible or difficult to mix in water. Miscible with ethanol, ether, chloroform and acetone.

Notes
Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 103-50-4
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.265
MDL Number MFCD00004780
InChI Key MHDVGSVTJDSBDK-UHFFFAOYSA-N
Synonym dibenzyl ether, benzyl ether, benzyl oxide, dibenzylether, oxybis methylene dibenzene, plastikator ba, ba plasticizer, benzylether, ether, dibenzyl, plasticator ba
PubChem CID 7657
ChEBI CHEBI:87411
IUPAC Name phenylmethoxymethylbenzene
SMILES C1=CC=C(C=C1)COCC2=CC=CC=C2

Specifications

Melting Point 3°C to 4°C
Density 1.04
Boiling Point 298°C
Flash Point 135°C (275°F)
Refractive Index 1.562
Quantity 250 g
UN Number UN3082
Beilstein 1911156
Merck Index 14,1132
Solubility Information Not miscible or difficult to mix in water. Miscible with ethanol,ether,chloroform and acetone.
Formula Weight 198.27
Percent Purity ≥98%
Chemical Name or Material Dibenzyl ether
Show More Show Less
Hazard Category H317
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 203-118-2
RTECSNumber DQ6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only