Learn More
Dibenzoyl peroxide, 97% (dry wt.), wet with 25% water
CAS: 94-36-0 | C14H10O4 | 242.23 g/mol
Supplier: Thermo Scientific Chemicals L131740B
Description
Benzoyl peroxide is widely utilized as a radical initiator to induce polymerizations. It finds applications for acne treatment, for bleaching flour, hair and teeth and for cross-linking polyester resins. It also has major applications in antiseptic and bleaching properties. It serves as a catalyst for polyester thermoset resins and as a hardener to start the polymerization process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzoyl peroxide is widely utilized as a radical initiator to induce polymerizations. It finds applications for acne treatment, for bleaching flour, hair and teeth and for cross-linking polyester resins. It also has major applications in antiseptic and bleaching properties. It serves as a catalyst for polyester thermoset resins and as a hardener to start the polymerization process.
Solubility
Soluble in ether and chloroform. Slightlysoluble in ethanol. Insoluble in water.
Notes
Incompatible with strong oxidizer, reducing agents, acids, bases, alcohols and metals.
Specifications
| Benzoyl peroxide | |
| (dry wt.), wet with 25% water | |
| MFCD00003071 | |
| UN3104 | |
| 14,1116 | |
| Soluble in ether and chloroform. Slightlysoluble in ethanol. Insoluble in water. | |
| C1=CC=C(C=C1)C(=O)OOC(=O)C2=CC=CC=C2 | |
| 242.23 | |
| CHEBI:82405 | |
| 97% |
| 94-36-0 | |
| C14H10O4 | |
| 1000 g | |
| 984320 | |
| benzoyl peroxide, dibenzoyl peroxide, peroxide, dibenzoyl, benzoperoxide, benzoyl superoxide, acetoxyl, lucidol, benoxyl, panoxyl, benzol peroxide | |
| OMPJBNCRMGITSC-UHFFFAOYSA-N | |
| benzoyl benzenecarboperoxoate | |
| 7187 | |
| 242.23 |
Chemical Identifiers
| 94-36-0 | |
| 242.23 | |
| OMPJBNCRMGITSC-UHFFFAOYSA-N | |
| 7187 | |
| benzoyl benzenecarboperoxoate |
| C14H10O4 | |
| MFCD00003071 | |
| benzoyl peroxide, dibenzoyl peroxide, peroxide, dibenzoyl, benzoperoxide, benzoyl superoxide, acetoxyl, lucidol, benoxyl, panoxyl, benzol peroxide | |
| CHEBI:82405 | |
| C1=CC=C(C=C1)C(=O)OOC(=O)C2=CC=CC=C2 |
Safety and Handling
GHS H Statement
H242-H319-H317
Heating may cause a fire.
Causes serious eye irritation.
May cause an allergic skin reaction.
P210-P220-P234-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P363-P501c
H242-H317-H319-H335
DOTInformation : Hazard Class: 5.2
EINECSNumber : 202-327-6
RTECSNumber : DM8575000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only