missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dibenzoyl-D-tartaric Acid Monohydrate 98.0+%, TCI America™

Catalog No. D139825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D139825G 25 g
D1398500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D139825G Supplier TCI America Supplier No. D139825G

Chemical Identifiers

CAS 80822-15-7
Molecular Formula C18H16O9
Molecular Weight (g/mol) 376.32
MDL Number MFCD00150722
InChI Key DXDIHODZARUBLA-DZWQFRRJNA-N
Synonym 2s,3s-2,3-bis benzoyloxy succinic acid hydrate, +-dibenzoyl-d-tartaric acid monohydrate, d-dbta, dibenzoyl-d-+-tartaric acid monohydrate, o,o'-dibenzoyl d-tartaric acid monohydrate, +-2,3-dibenzoyl-d-tartaric acid monohydrate, +-dibenzoyl-d-tartaric acid, 2s,3s-+-dibenzoyl-d-tartaric acid monohydrate, 2s,3s-+-o,o'-dibenzoyltartaric acid monohydrate, butanedioic acid, 2,3-bis benzoyloxy-, monohydrate, 2s,3s
PubChem CID 11667949
IUPAC Name (2S,3S)-2,3-bis(benzoyloxy)butanedioic acid hydrate
SMILES O.OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 156°C
Color White
Quantity 25 g
Formula Weight 358.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Dibenzoyl-D-tartaric Acid Monohydrate