missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dibenzoyl-D-tartaric acid monohydrate, 99%

Catalog No. AAA1650314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1650314 25 g
AAA1650322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1650314 Supplier Thermo Scientific Chemicals Supplier No. A1650314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 80822-15-7 | C18H16O9 | 376.32 g/mol

(+)-Dibenzoyl-D-tartaric acid monohydrate is used as a chiral separation and racemization agent for amines. It is also used as a pharmaceutical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-Dibenzoyl-D-tartaric acid monohydrate is used as a chiral separation and racemization agent for amines. It is also used as a pharmaceutical raw material.

Solubility
Soluble in methanol. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 80822-15-7
Molecular Formula C18H16O9
Molecular Weight (g/mol) 376.32
MDL Number MFCD00150722
InChI Key DXDIHODZARUBLA-DZWQFRRJNA-N
Synonym 2s,3s-2,3-bis benzoyloxy succinic acid hydrate, +-dibenzoyl-d-tartaric acid monohydrate, d-dbta, dibenzoyl-d-+-tartaric acid monohydrate, o,o'-dibenzoyl d-tartaric acid monohydrate, +-2,3-dibenzoyl-d-tartaric acid monohydrate, +-dibenzoyl-d-tartaric acid, 2s,3s-+-dibenzoyl-d-tartaric acid monohydrate, 2s,3s-+-o,o'-dibenzoyltartaric acid monohydrate, butanedioic acid, 2,3-bis benzoyloxy-, monohydrate, 2s,3s
PubChem CID 11667949
SMILES O.OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 87°C to 91°C
Quantity 25 g
Optical Rotation +109° (c=2 in Ethanol)
Beilstein 6121616
Solubility Information Soluble in methanol. Slightly soluble in water.
IUPAC Name (2S,3S)-2,3-dibenzoyloxybutanedioic acid;hydrate
Formula Weight 376.33 (358.31 Anhydrous)
Percent Purity 99%
Chemical Name or Material (+)-Dibenzoyl-D-tartaric acid monohydrate
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 241-097-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only