missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo[b,def]chrysene 98.0+%, TCI America™

Catalog No. D1005100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1005100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1005100MG Supplier TCI America Supplier No. D1005100MG

Chemical Identifiers

CAS 189-64-0
Molecular Formula C24H14
Molecular Weight (g/mol) 302.376
MDL Number MFCD00039579
InChI Key RXUSYFJGDZFVND-UHFFFAOYSA-N
Synonym dibenzo b,def chrysene, dibenzo a,h pyrene, db a,h p, 3,4,8,9-dibenzpyrene, 1,2,6,7-dibenzopyrene, 3,4,8,9-dibenzopyrene, unii-xr5r3v8bjk, 3,4:8,9-dibenzopyrene
PubChem CID 9108
ChEBI CHEBI:82313
SMILES C1=CC=C2C3=C4C(=CC2=C1)C=CC5=C4C(=CC6=CC=CC=C56)C=C3

Specifications

Melting Point 308°C
Boiling Point 596°C
Quantity 100 mg
Formula Weight 302.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzo[b,def]chrysene
RTECSNumber HO5775000
TSCA No