CAS RN 189-64-0
IUPAC Name:
hexacyclo[10.10.2.0²,⁷.0⁹,²³.0¹³,¹⁸.0²⁰,²⁴]tetracosa-1(23),2,4,6,8,10,12(24),13,15,17,19,21-dodecaene
Synonyms:
dibenzo[a,h]pyrene
Molecular Weight (g/mol):
302.38
Molecular Formula:
C24H14
InChi Key:
RXUSYFJGDZFVND-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(=C1)C=C1C=CC3=C4C(C=CC2=C14)=CC1=CC=CC=C31
1
–
1
of
1
results
1
Dibenzo[b,def]chrysene 98.0+%, TCI America™
CAS: 189-64-0 Molecular Formula: C24H14 Molecular Weight (g/mol): 302.376 MDL Number: MFCD00039579 InChI Key: RXUSYFJGDZFVND-UHFFFAOYSA-N Synonym: dibenzo b,def chrysene,dibenzo a,h pyrene,db a,h p,3,4,8,9-dibenzpyrene,1,2,6,7-dibenzopyrene,3,4,8,9-dibenzopyrene,unii-xr5r3v8bjk,3,4:8,9-dibenzopyrene PubChem CID: 9108 ChEBI: CHEBI:82313 SMILES: C1=CC=C2C3=C4C(=CC2=C1)C=CC5=C4C(=CC6=CC=CC=C56)C=C3