missing translation for 'onlineSavingsMsg'
Learn More

Dibenz[a,h]anthracene 98.0+%, TCI America™

Catalog No. D0145100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0145100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D0145100MG Supplier TCI America Supplier No. D0145100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 53-70-3
Molecular Formula C22H14
Molecular Weight (g/mol) 278.354
MDL Number MFCD00003708
InChI Key LHRCREOYAASXPZ-UHFFFAOYSA-N
Synonym Naphtho[1′C,2′C:2,3]phenanthrene
PubChem CID 5889
ChEBI CHEBI:35299
IUPAC Name naphtho[1,2-b]phenanthrene
SMILES C1=CC=C2C(=C1)C=CC3=CC4=C(C=CC5=CC=CC=C54)C=C32

Specifications

Physical Form Powder/Crystals
UN Number 3077
Color Yellow
Melting Point 266°C
Boiling Point 524°C
Quantity 100 mg
Formula Weight 278.35
Percent Purity ≥98.0% (GC)
Chemical Name or Material Dibenz[a,h]anthracene
RTECSNumber HN2625000
TSCA Yes