missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibasic Sodium Phosphate, Heptahydrate, Crystal, USP, 98-100.5%, Spectrum™ Chemical
SO140, 7782-85-6, Na2HPO4·7H2O
$616.00 - $11464.83
Chemical Identifiers
| CAS | 7782-85-6 |
|---|---|
| Molecular Formula | H15Na2O11P |
| Molecular Weight (g/mol) | 268.06 |
| MDL Number | MFCD00149180 |
| InChI Key | LVXHNCUCBXIIPE-UHFFFAOYSA-L |
| IUPAC Name | disodium phosphoric acid heptahydrate |
| SMILES | O.[Na+].[Na+].OP([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602159
|
Spectrum Chemical Mfg Cor
SO140500GM |
500 g | Poly Bottle |
Each for $616.00
|
|
||||
|
18602160
|
Spectrum Chemical Mfg Cor
SO14025KG |
2.5 kg | Poly Bottle |
Each for $1,197.43
|
|
||||
|
18602161
|
Spectrum Chemical Mfg Cor
SO14012KG |
12 kg | Poly Pail |
Each for $3,437.67
|
|
||||
|
18602162
|
Spectrum Chemical Mfg Cor
SO14050KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Dibasic Sodium Phosphate, Heptahydrate, Crystal, USP is used as a saline laxative to treat constipation or to clean the bowel before a colonoscopy. Spectrum Chemical USP products, including all USP Sodium Phosphate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 7782-85-6 | |
| 268.06 | |
| LVXHNCUCBXIIPE-UHFFFAOYSA-L | |
| O.[Na+].[Na+].OP([O-])([O-])=O |
| H15Na2O11P | |
| MFCD00149180 | |
| disodium phosphoric acid heptahydrate |
Specifications
| 7782-85-6 | |
| 500 g | |
| MFCD00149180 | |
| O.[Na+].[Na+].OP([O-])([O-])=O | |
| 268.06 | |
| USP | |
| Poly Bottle |
| 100% | |
| H15Na2O11P | |
| LVXHNCUCBXIIPE-UHFFFAOYSA-L | |
| disodium phosphoric acid heptahydrate | |
| 98 to 100.5% | |
| 43.0-50% |