missing translation for 'onlineSavingsMsg'
Learn More

Dibasic Sodium Phosphate, Heptahydrate, Crystal, USP, 98-100.5%, Spectrum™ Chemical

Catalog No. 18602162
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602162 50 kg Poly Bottle
18602159 500 g Poly Bottle
18602160 2.5 kg Poly Bottle
18602161 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602162 Supplier Spectrum Chemical Mfg Cor Supplier No. SO14050KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SO140, 7782-85-6, Na2HPO4·7H2O

Spectrum™ Chemical Dibasic Sodium Phosphate, Heptahydrate, Crystal, USP is used as a saline laxative to treat constipation or to clean the bowel before a colonoscopy. Spectrum Chemical USP products, including all USP Sodium Phosphate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H17Na2O11P
Molecular Weight (g/mol) 270.08
MDL Number MFCD00149180
InChI Key PYLIXCKOHOHGKQ-UHFFFAOYSA-N
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O

Specifications

CAS Max % 100%
Physical Form Crystals or Needles
Quantity 50 kg
Percent Purity 98 to 100.5%
Grade USP
Loss on Drying 43.0-50%
Packaging Poly Bottle
Chemical Name or Material Dibasic Sodium Phosphate, Heptahydrate, Crystal