missing translation for 'onlineSavingsMsg'
Learn More

Dibasic Sodium Phosphate, Heptahydrate, Crystal, USP, 98-100.5%, Spectrum™ Chemical

Catalog No. 18602159
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602159 500 g Poly Bottle
18602160 2.5 kg Poly Bottle
18602161 12 kg Poly Pail
18602162 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602159 Supplier Spectrum Chemical Mfg Cor Supplier No. SO140500GM

SO140, 7782-85-6, Na2HPO4·7H2O

Spectrum™ Chemical Dibasic Sodium Phosphate, Heptahydrate, Crystal, USP is used as a saline laxative to treat constipation or to clean the bowel before a colonoscopy. Spectrum Chemical USP products, including all USP Sodium Phosphate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
MDL Number MFCD00149180
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 98 to 100.5%
Grade USP
Loss on Drying 43.0-50%
Packaging Poly Bottle