missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibasic Sodium Phosphate, Dried, USP, 98-100.5%, Spectrum™ Chemical
SO138, 7558-79-4, Na2HPO4
$608.63 - $10122.47
Chemical Identifiers
| CAS | 7558-79-4 |
|---|---|
| Molecular Formula | HNa2O4P |
| Molecular Weight (g/mol) | 141.96 |
| MDL Number | MFCD00003496 |
| InChI Key | BNIILDVGGAEEIG-UHFFFAOYSA-L |
| IUPAC Name | disodium hydrogen phosphate |
| SMILES | [Na+].[Na+].OP([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602152
|
Spectrum Chemical Mfg Cor
SO138500GMPL |
500 g | Poly Bottle |
Each for $608.63
|
|
||||
|
18602154
|
Spectrum Chemical Mfg Cor
SO13825KGPL |
2.5 kg | Poly Bottle |
Each for $1,874.53
|
|
||||
|
18602153
|
Spectrum Chemical Mfg Cor
SO138500GMPLCS6 |
6 x 500 g | Poly Bottle |
Case of 6 Each for $2,553.93
|
|
||||
|
18602155
|
Spectrum Chemical Mfg Cor
SO13825KGPLCS4 |
4 x 2.5 kg | Poly Bottle |
Case of 4 Each for $6,480.43
|
|
||||
|
18602156
|
Spectrum Chemical Mfg Cor
SO13812KGPL |
12 kg | Poly Pail |
Each for $4,432.73
|
|
||||
|
18602157
|
Spectrum Chemical Mfg Cor
SO13825KGBL |
25 kg | Fiber Drum |
Each for $7,830.63
|
|
||||
|
18602158
|
Spectrum Chemical Mfg Cor
SO13850KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Dibasic Sodium Phosphate, Dried, USP is used as a saline laxative to treat constipation or to clean the bowel before a colonoscopy. Spectrum Chemical USP products, including all USP Sodium Phosphate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 7558-79-4 | |
| 141.96 | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| [Na+].[Na+].OP([O-])([O-])=O |
| HNa2O4P | |
| MFCD00003496 | |
| disodium hydrogen phosphate |
Specifications
| 7558-79-4 | |
| 500 g | |
| MFCD00003496 | |
| [Na+].[Na+].OP([O-])([O-])=O | |
| 141.96 | |
| USP | |
| Poly Bottle |
| 100% | |
| HNa2O4P | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| disodium hydrogen phosphate | |
| 98 to 100.5% | |
| 5% |