missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibasic Sodium Phosphate, Dried, USP, 98-100.5%, Spectrum™ Chemical
SO138, 7558-79-4, Na2HPO4
Supplier: Spectrum Chemical Mfg Cor SO13812KGPL
Description
Spectrum™ Chemical Dibasic Sodium Phosphate, Dried, USP is used as a saline laxative to treat constipation or to clean the bowel before a colonoscopy. Spectrum Chemical USP products, including all USP Sodium Phosphate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 7558-79-4 | |
| 12 kg | |
| MFCD00003496 | |
| [Na+].[Na+].OP([O-])([O-])=O | |
| 141.96 | |
| USP | |
| Poly Pail |
| 100% | |
| HNa2O4P | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| disodium hydrogen phosphate | |
| 98 to 100.5% | |
| 5% |
Chemical Identifiers
| 7558-79-4 | |
| 141.96 | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| [Na+].[Na+].OP([O-])([O-])=O |
| HNa2O4P | |
| MFCD00003496 | |
| disodium hydrogen phosphate |