missing translation for 'onlineSavingsMsg'
Learn More

Diazoxide 98.0+%, TCI America™

Supplier:  TCI America D5402250MG

 View more versions of this product

Catalog No. D5402250MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Diazoxide
332°C
C8H7ClN2O2S
250 mg
GDLBFKVLRPITMI-UHFFFAOYSA-N
7-chloro-3-methyl-4H-1λ⁶,2,4-benzothiadiazine-1,1-dione
3019
230.67
Crystalline Powder
364-98-7
White-Yellow
MFCD00078578
diazoxide, hyperstat, proglycem, hypertonalum, eudemine, proglicem, dizoxide, mutabase, diazossido, 7-chloro-3-methyl-2h-1,2,4-benzothiadiazine 1,1-dioxide
CC1=NS(=O)(=O)C2=CC(Cl)=CC=C2N1
230.67
CHEBI:4495
≥98.0% (HPLC,T)

Chemical Identifiers

364-98-7
230.67
GDLBFKVLRPITMI-UHFFFAOYSA-N
3019
7-chloro-3-methyl-4H-1λ⁶,2,4-benzothiadiazine-1,1-dione
C8H7ClN2O2S
MFCD00078578
diazoxide, hyperstat, proglycem, hypertonalum, eudemine, proglicem, dizoxide, mutabase, diazossido, 7-chloro-3-methyl-2h-1,2,4-benzothiadiazine 1,1-dioxide
CHEBI:4495
CC1=NS(=O)(=O)C2=CC(Cl)=CC=C2N1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DK8185000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions