missing translation for 'onlineSavingsMsg'
Learn More

Diazoxide 98.0+%, TCI America™

Catalog No. p-7128757
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5402250MG 250 mg
D54021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5402250MG Supplier TCI America Supplier No. D5402250MG

Chemical Identifiers

CAS 364-98-7
Molecular Formula C8H7ClN2O2S
Molecular Weight (g/mol) 230.67
MDL Number MFCD00078578
InChI Key GDLBFKVLRPITMI-UHFFFAOYSA-N
Synonym diazoxide, hyperstat, proglycem, hypertonalum, eudemine, proglicem, dizoxide, mutabase, diazossido, 7-chloro-3-methyl-2h-1,2,4-benzothiadiazine 1,1-dioxide
PubChem CID 3019
ChEBI CHEBI:4495
IUPAC Name 7-chloro-3-methyl-4H-1λ⁶,2,4-benzothiadiazine-1,1-dione
SMILES CC1=NS(=O)(=O)C2=CC(Cl)=CC=C2N1

Specifications

Melting Point 332°C
Color White-Yellow
Quantity 250 mg
Formula Weight 230.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diazoxide
RTECSNumber DK8185000
TSCA Yes