missing translation for 'onlineSavingsMsg'
Learn More

Diazinon, SPEX CertiPrep™

Catalog No. s1175
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S1175 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S1175 Supplier SPEX CertiPrep Group LLC Supplier No. S1175

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Diazinon333-41-51000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 333-41-5 , 67-64-1
Molecular Formula C12H21N2O3PS
Molecular Weight (g/mol) 304.345
InChI Key FHIVAFMUCKRCQO-UHFFFAOYSA-N
PubChem CID 3017
ChEBI CHEBI:34682
IUPAC Name diethoxy-(6-methyl-2-propan-2-ylpyrimidin-4-yl)oxy-sulfanylidene-$l^{5}-phosphane
SMILES CCOP(=S)(OCC)OC1=NC(=NC(=C1)C)C(C)C

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Methods For US EPA Method 622
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Acetone
Quantity 1 mL
Chemical Name or Material Diazinon
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only