missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diallyl Phthalate 98.0+%, TCI America™
$41.02 - $78.29
Chemical Identifiers
| CAS | 131-17-9 |
|---|---|
| Molecular Formula | C14H14O4 |
| Molecular Weight (g/mol) | 246.262 |
| MDL Number | MFCD00008646 |
| InChI Key | QUDWYFHPNIMBFC-UHFFFAOYSA-N |
| Synonym | diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st |
| PubChem CID | 8560 |
| IUPAC Name | bis(prop-2-enyl) benzene-1,2-dicarboxylate |
| SMILES | C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P029025ML
|
TCI America
P029025ML |
25 mL |
Each for $41.02
|
|
|||||
|
P0290500ML
|
TCI America
P0290500ML |
500 mL |
Each for $78.29
|
|
|||||
Chemical Identifiers
| 131-17-9 | |
| 246.262 | |
| QUDWYFHPNIMBFC-UHFFFAOYSA-N | |
| 8560 | |
| C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C |
| C14H14O4 | |
| MFCD00008646 | |
| diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st | |
| bis(prop-2-enyl) benzene-1,2-dicarboxylate |
Specifications
| 131-17-9 | |
| 158°C | |
| MFCD00008646 | |
| 3082 | |
| QUDWYFHPNIMBFC-UHFFFAOYSA-N | |
| bis(prop-2-enyl) benzene-1,2-dicarboxylate | |
| 8560 | |
| ≥98.0% (GC) | |
| Diallyl Phthalate |
| Yellow | |
| C14H14O4 | |
| 25 mL | |
| diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st | |
| C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C | |
| 246.262 | |
| 246.26 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-1325
RTECSNumber : CZ4200000
TSCA : Yes