missing translation for 'onlineSavingsMsg'
Learn More

Diallyl phthalate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 276620025

Quantity 2.5L
Packaging Glass bottle

 View more versions of this product

Catalog No. AC276620025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

131-17-9
246.26
QUDWYFHPNIMBFC-UHFFFAOYSA-N
8560
C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C
C14H14O4
MFCD00008646
diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st
bis(prop-2-enyl) benzene-1,2-dicarboxylate

Specifications

Diallyl phthalate
1.1200g/mL
166°C
97.5% min. (GC)
C14H14O4
2.5L
3082
1.12
Solubility in water: 148mg/l (20°C)
C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C
246.26
13 mPa.s (20°C)
98%
131-17-9
300.0°C
Authentic
Glass bottle
1.5180 to 1.5200
MFCD00008646
09, 4120
diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st
QUDWYFHPNIMBFC-UHFFFAOYSA-N
bis(prop-2-enyl) benzene-1,2-dicarboxylate
8560
246.26
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Suspected of causing genetic defects.
May cause damage to organs thr

GHS P Statement
Avoid release to the environment.
Do not get in eyes,on skin,or on clothing.
Call a POISON CENTRE or doctor/physician.

GHS Signal Word: Warning

EINECSNumber : 205-016-3

SDS
Documents

Documents

Promotions

Promotions