missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 541-16-2 |
|---|---|
| Molecular Formula | C11H20O4 |
| Molecular Weight (g/mol) | 216.28 |
| MDL Number | MFCD00008810 |
| InChI Key | CLPHAYNBNTVRDI-UHFFFAOYSA-N |
| Synonym | di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate |
| PubChem CID | 68324 |
| IUPAC Name | ditert-butyl propanedioate |
| SMILES | CC(C)(C)OC(=O)CC(=O)OC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC406620100
|
N/A | 10g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC406620500
|
N/A | 50g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C11H20O4 | |
| MFCD00008810 | |
| di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate | |
| ditert-butyl propanedioate |
Specifications
| 541-16-2 | |
| 110°C to 111°C (22.0mmHg) | |
| Authentic | |
| Glass bottle | |
| CH2(CO2C(CH3)3)2 | |
| MFCD00008810 | |
| 01,210 | |
| 15, 3041 | |
| CLPHAYNBNTVRDI-UHFFFAOYSA-N | |
| ditert-butyl propanedioate | |
| 68324 | |
| 98% |
| 0.9660g/mL | |
| 88°C | |
| 97.5% min. (GC) | |
| C11H20O4 | |
| 1.4175 to 1.4195 | |
| 02, III, 1621 | |
| 0.966 | |
| di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate | |
| CC(C)(C)OC(=O)CC(=O)OC(C)(C)C | |
| 216.28 | |
| 216.28 | |
| Di-tert-butyl malonate |
Safety and Handling
EINECSNumber : 208-769-6
RUO – Research Use Only