missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl malonate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 406620500

Quantity 50g

 View more versions of this product

Catalog No. AC406620500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

541-16-2
216.28
CLPHAYNBNTVRDI-UHFFFAOYSA-N
68324
CC(C)(C)OC(=O)CC(=O)OC(C)(C)C
C11H20O4
MFCD00008810
di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate
ditert-butyl propanedioate

Specifications

Di-tert-butyl malonate
0.9660g/mL
88°C
97.5% min. (GC)
C11H20O4
CH2(CO2C(CH3)3)2
MFCD00008810
01,210
15, 3041
CLPHAYNBNTVRDI-UHFFFAOYSA-N
ditert-butyl propanedioate
68324
98%
541-16-2
110°C to 111°C (22.0mmHg)
Authentic
Glass bottle
1.4175 to 1.4195
50g
02, III, 1621
0.966
di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate
CC(C)(C)OC(=O)CC(=O)OC(C)(C)C
216.28
216.28
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-769-6

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...