missing translation for 'onlineSavingsMsg'
Learn More

Di-p-tolyl Disulfide 98.0+%, TCI America™

Catalog No. p-7136594
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T107425G 25 g
T1074100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T107425G Supplier TCI America Supplier No. T107425G
Only null left

Chemical Identifiers

CAS 103-19-5
Molecular Formula C14H14S2
Molecular Weight (g/mol) 246.386
MDL Number MFCD00008547
InChI Key TZOVOULUMXXLOJ-UHFFFAOYSA-N
Synonym p-tolyl disulfide, 1,2-di-p-tolyldisulfane, di-p-tolyl disulfide, disulfide, bis 4-methylphenyl, biodylon, kresulfin, 4-methylphenyl disulfide, bis 4-methylphenyl disulfide, p-tolyldisulfide, di-4-tolyl disulfide
PubChem CID 66027
IUPAC Name 1-methyl-4-[(4-methylphenyl)disulfanyl]benzene
SMILES CC1=CC=C(C=C1)SSC2=CC=C(C=C2)C

Specifications

Melting Point 45°C
Quantity 25 g
Formula Weight 246.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Di-p-tolyl Disulfide
EINECSNumber (3)-1126
RTECSNumber JO1526250
TSCA Yes