accessibility menu, dialog, popup

UserName

CAS RN 103-19-5

IUPAC Name: 1-methyl-4-[(4-methylphenyl)disulfanyl]benzene
Synonyms: disulfide, bis(4-methylphenyl)
Molecular Weight (g/mol): 246.39
Molecular Formula: C14H14S2
InChi Key: TZOVOULUMXXLOJ-UHFFFAOYSA-N
SMILES: CC1=CC=C(SSC2=CC=C(C)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results