missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-p-toluoyl-D-tartaric acid, 98%
CAS: 32634-68-7 | C20H18O8 | 386.36 g/mol
$540.18 - $540.18
Chemical Identifiers
| CAS | 32634-68-7 |
|---|---|
| Molecular Formula | C20H18O8 |
| Molecular Weight (g/mol) | 386.36 |
| MDL Number | MFCD00008552 |
| InChI Key | CMIBUZBMZCBCAT-HOTGVXAUSA-N |
| Synonym | di-p-toluoyl-d-tartaric acid, +-di-1,4-toluoyl-d-tartaric acid, 2s,3s-2,3-bis 4-methylbenzoyl oxy succinic acid, unii-4480ma5qip, ditoluoyltartaric acid, +, +-dtta, butanedioic acid, 2,3-bis 4-methylbenzoyl oxy-, 2s,3s, 2s,3s-2,3-bis 4-methylbenzoyl oxy butanedioic acid, --di-p-toluoyl-l-trataric acid |
| PubChem CID | 263211 |
| SMILES | CC1=CC=C(C=C1)C(=O)O[C@@H]([C@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1322322
|
Thermo Scientific Chemicals
L1322322 |
100 g |
Each for $540.18
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 32634-68-7 | |
| 386.36 | |
| CMIBUZBMZCBCAT-HOTGVXAUSA-N | |
| 263211 |
| C20H18O8 | |
| MFCD00008552 | |
| di-p-toluoyl-d-tartaric acid, +-di-1,4-toluoyl-d-tartaric acid, 2s,3s-2,3-bis 4-methylbenzoyl oxy succinic acid, unii-4480ma5qip, ditoluoyltartaric acid, +, +-dtta, butanedioic acid, 2,3-bis 4-methylbenzoyl oxy-, 2s,3s, 2s,3s-2,3-bis 4-methylbenzoyl oxy butanedioic acid, --di-p-toluoyl-l-trataric acid | |
| CC1=CC=C(C=C1)C(=O)O[C@@H]([C@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O |
Specifications
| 169°C to 171°C | |
| C20H18O8 | |
| 3225584 | |
| CMIBUZBMZCBCAT-HOTGVXAUSA-N | |
| (2S,3S)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid | |
| 263211 | |
| 98% | |
| +139° (c=1 in Ethanol) |
| 32634-68-7 | |
| MFCD00008552 | |
| di-p-toluoyl-d-tartaric acid, +-di-1,4-toluoyl-d-tartaric acid, 2s,3s-2,3-bis 4-methylbenzoyl oxy succinic acid, unii-4480ma5qip, ditoluoyltartaric acid, +, +-dtta, butanedioic acid, 2,3-bis 4-methylbenzoyl oxy-, 2s,3s, 2s,3s-2,3-bis 4-methylbenzoyl oxy butanedioic acid, --di-p-toluoyl-l-trataric acid | |
| CC1=CC=C(C=C1)C(=O)O[C@@H]([C@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O | |
| 386.36 | |
| 386.36 | |
| 100 g | |
| Di-p-toluoyl-D-tartaric acid |
Safety and Handling
EINECSNumber : 251-132-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only