missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di(N-succinimidyl) 3,3'-Dithiodipropionate 97.0+%, TCI America™
Supplier: TCI America D24735G
Specifications
| Di(N-succinimidyl) 3,3′-Dithiodipropionate [Cross-linking Reagent] | |
| 133°C | |
| MFCD00042045 | |
| 3,3′-Dithiodipropionic Acid Bis(N-succinimidyl Ester), 3,3′-Dithiodipropionic Acid Di(N-succinimidyl Ester), DTSP | |
| C1CC(=O)N(C1=O)OC(=O)CCSSCCC(=O)ON2C(=O)CCC2=O | |
| 404.408 | |
| 404.41 | |
| Crystalline Powder |
| 57757-57-0 | |
| C14H16N2O8S2 | |
| 5 g | |
| FXYPGCIGRDZWNR-UHFFFAOYSA-N | |
| (2,5-dioxopyrrolidin-1-yl) 3-[[3-(2,5-dioxopyrrolidin-1-yl)oxy-3-oxopropyl]disulfanyl]propanoate | |
| 93313 | |
| ≥97.0% (N) |
Chemical Identifiers
| 57757-57-0 | |
| 404.408 | |
| FXYPGCIGRDZWNR-UHFFFAOYSA-N | |
| (2,5-dioxopyrrolidin-1-yl) 3-[[3-(2,5-dioxopyrrolidin-1-yl)oxy-3-oxopropyl]disulfanyl]propanoate |
| C14H16N2O8S2 | |
| MFCD00042045 | |
| 93313 | |
| C1CC(=O)N(C1=O)OC(=O)CCSSCCC(=O)ON2C(=O)CCC2=O |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator