missing translation for 'onlineSavingsMsg'
Learn More

Di(N-succinimidyl) 3,3'-Dithiodipropionate 97.0+%, TCI America™

Catalog No. p-7127318
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2473200MG 200 mg
D24731G 1 g
D24735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D2473200MG Supplier TCI America Supplier No. D2473200MG
Only null left

Chemical Identifiers

CAS 57757-57-0
Molecular Formula C14H16N2O8S2
Molecular Weight (g/mol) 404.408
MDL Number MFCD00042045
InChI Key FXYPGCIGRDZWNR-UHFFFAOYSA-N
PubChem CID 93313
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-[[3-(2,5-dioxopyrrolidin-1-yl)oxy-3-oxopropyl]disulfanyl]propanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCSSCCC(=O)ON2C(=O)CCC2=O

Specifications

Melting Point 133°C
Quantity 200 mg
Synonym 3,3′C-Dithiodipropionic Acid Bis(N-succinimidyl Ester), 3,3′C-Dithiodipropionic Acid Di(N-succinimidyl Ester), DTSP
Formula Weight 404.41
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Di(N-succinimidyl) 3,3′-Dithiodipropionate [Cross-linking Reagent]
TSCA No
Recommended Storage Refrigerator